Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008445.2 | NP_032471.2 | 7 - 8 | 3146 | 96 | |
GenBank mRNA | AF013116.1 | - | 7 - 8 | 2722 | 96 | |
AK029381.1 | - | 1 - 2 | 546 | 96 | ||
AK139894.1 | - | 7 - 8 | 822 | 96 | ||
AK147572.1 | - | 7 - 8 | 2948 | 96 | ||
AK220283.1 | - | 8 - 9 | 2871 | 96 | ||
BC052732.1 | - | 7 - 8 | 2303 | 96 | ||
BC053053.1 | - | 7 - 8 | 3088 | 96 | ||
BC062648.1 | - | 7 - 8 | 2821 | 95 | ||
BC127063.1 | - | 7 - 8 | 2358 | 40109965 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |