Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gipr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080815.1 | NP_001074284.1 | 1 - 2 | 67 | 110 | |
GenBank mRNA | AK050472.1 | - | 1 - 2 | 68 | 110 | |
AK146552.1 | - | 1 - 2 | 68 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Mus musculus Probe spans exons |