Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190490.1 | NP_001177419.1 | 14 - 15 | 1724 | 129 | |
NM_032418.2 | NP_115794.1 | 13 - 14 | 1802 | 129 | ||
GenBank mRNA | BC075715.1 | - | 14 - 15 | 1632 | 4989259 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |