Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039106.3 | NP_001034195.2 | 5 - 6 | 1481 | 71 | |
NM_001042719.2 | NP_001036184.1 | 6 - 7 | 1583 | 71 | ||
NM_001284399.1 | NP_001271328.1 | 7 - 8 | 1555 | 71 | ||
NM_176845.5 | NP_789815.3 | 5 - 6 | 1481 | 71 | ||
GenBank mRNA | AK014975.1 | - | 3 - 4 | 331 | 71 | |
AK048690.1 | - | 6 - 7 | 1041 | 71 | ||
AK129425.1 | - | 3 - 4 | 892 | 71 | ||
AK155294.1 | - | 7 - 8 | 1512 | 71 | ||
AK170271.1 | - | 6 - 7 | 1195 | 71 | ||
BC043475.1 | - | 4 - 5 | 735 | 5365658 | 71 | |
CF532335.1 | - | 4 - 5 | 403 | 71 | ||
DQ272481.1 | - | 5 - 6 | 1480 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |