Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011074.2 | NP_035204.2 | 2 - 3 | 244 | 67 | |
GenBank mRNA | AF033655.1 | - | 2 - 3 | 203 | 67 | |
AK016370.1 | - | 2 - 3 | 186 | 67 | ||
AK045083.1 | - | 2 - 3 | 245 | 67 | ||
AK051283.1 | - | 2 - 3 | 218 | 67 | ||
AK051411.1 | - | 2 - 3 | 187 | 67 | ||
AK083790.1 | - | 2 - 3 | 187 | 67 | ||
AK129226.1 | - | 2 - 3 | 71 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mus musculus Probe spans exons |