Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500000388.mm.1 |
TC0500000048.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500000388.mm.1 |
NM_001289595.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000388.mm.1 |
NM_001289596.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000388.mm.1 |
NM_178766.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000393.mm.1 |
TC0500000048.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500000393.mm.1 |
NM_001289595.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000393.mm.1 |
NM_001289596.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000393.mm.1 |
NM_178766.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000385.mm.1 |
TC0500000048.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500000385.mm.1 |
NM_001289595.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000385.mm.1 |
NM_001289596.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000385.mm.1 |
NM_178766.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000387.mm.1 |
TC0500000048.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500000387.mm.1 |
NM_001289596.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000387.mm.1 |
NM_178766.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000391.mm.1 |
TC0500000048.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500000391.mm.1 |
NM_001289595.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000391.mm.1 |
NM_001289596.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000391.mm.1 |
NM_178766.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000394.mm.1 |
TC0500000048.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500000394.mm.1 |
NM_001289595.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000394.mm.1 |
NM_001289596.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000394.mm.1 |
NM_178766.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000386.mm.1 |
TC0500000048.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500000386.mm.1 |
NM_001289595.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000386.mm.1 |
NM_001289596.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000386.mm.1 |
NM_178766.5 |
Refseq |