Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300011642.mm.1 |
NM_001289492.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011642.mm.1 |
NM_018734.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011642.mm.1 |
NM_018734.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011642.mm.1 |
TC0300001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300011646.mm.1 |
NM_001289492.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011646.mm.1 |
NM_018734.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011646.mm.1 |
NM_018734.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011646.mm.1 |
TC0300001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300011653.mm.1 |
NM_001289492.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011653.mm.1 |
NM_001289493.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011653.mm.1 |
NM_018734.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011653.mm.1 |
NM_018734.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011653.mm.1 |
TC0300001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300011650.mm.1 |
NM_001289492.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011650.mm.1 |
TC0300001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300011654.mm.1 |
NM_001289492.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011654.mm.1 |
NM_001289493.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011654.mm.1 |
NM_018734.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011654.mm.1 |
NM_018734.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011654.mm.1 |
TC0300001446.mm.1 |
MTA TC |