Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163522.1 | NP_001156994.1 | 4 - 5 | 549 | 100 | |
GenBank mRNA | AB034693.1 | - | 4 - 5 | 502 | 100 | |
AK029348.1 | - | 516 | 100 | |||
BC003706.1 | - | 4 - 5 | 510 | 3485788 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |