Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp3cc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK015002.1 | - | 3 - 4 | 596 | 109 | |
AK133393.1 | - | 3 - 4 | 617 | 109 | ||
BC046617.1 | - | 3 - 4 | 422 | 109 | ||
BC141079.1 | - | 3 - 4 | 437 | 9055691 | 109 | |
M81475.1 | - | 3 - 4 | 590 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |