Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199003.1 | NP_001185932.1 | 7 - 8 | 787 | 76 | |
NM_011880.3 | NP_036010.2 | 7 - 8 | 787 | 76 | ||
GenBank mRNA | AF011360.1 | - | 7 - 8 | 632 | 76 | |
BC051133.1 | - | 7 - 8 | 742 | 6588901 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |