Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrriq3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028938.2 | NP_083214.2 | 3 - 4 | 812 | 129 | |
GenBank mRNA | AK015333.1 | - | 3 - 4 | 709 | 129 | |
AK015758.1 | - | 3 - 4 | 659 | 129 | ||
AK016634.1 | - | 3 - 4 | 707 | 129 | ||
AK132656.1 | - | 3 - 4 | 777 | 129 | ||
BC112424.1 | - | 3 - 4 | 669 | 40062088 | 129 | |
BC113130.1 | - | 3 - 4 | 669 | 40062093 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |