Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024645.1 | NP_001019816.1 | 21 - 22 | 1947 | 64 | |
GenBank mRNA | AK131164.1 | - | 21 - 22 | 1890 | 64 | |
AK148355.1 | - | 21 - 22 | 1970 | 64 | ||
AK158466.1 | - | 22 - 23 | 2193 | 64 | ||
BC036313.1 | - | 6 - 7 | 509 | 64 | ||
BC060252.1 | - | 2 - 3 | 487 | 64 | ||
BC082805.1 | - | 21 - 22 | 1867 | 64 | ||
BC092223.1 | - | 21 - 22 | 1865 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |