Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136057.2 | NP_001129529.1 | 12 - 13 | 1399 | 114 | |
NM_001146183.1 | NP_001139655.1 | 12 - 13 | 1376 | 114 | ||
NM_009947.3 | NP_034077.1 | 12 - 13 | 1202 | 114 | ||
GenBank mRNA | AB008893.1 | - | 12 - 13 | 1202 | 114 | |
AK134548.1 | - | 12 - 13 | 1221 | 114 | ||
AK144410.1 | - | 12 - 13 | 1228 | 114 | ||
BC050766.1 | - | 12 - 13 | 1224 | 6591063 | 114 | |
BC051994.1 | - | 10 - 11 | 1856 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |