Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF068707.1 | - | 15 - 16 | 1647 | 87 | |
AK147177.1 | - | 16 - 17 | 1686 | 87 | ||
AK151844.1 | - | 16 - 17 | 1678 | 87 | ||
AK194854.1 | - | 1 - 2 | 116 | 87 | ||
BC058402.1 | - | 14 - 15 | 1692 | 87 | ||
BC080798.1 | - | 15 - 16 | 1594 | 87 | ||
BC108374.1 | - | 16 - 17 | 1673 | 4936738 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |