Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rec8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020002.3 | NP_064386.2 | 4 - 5 | 510 | 87 | |
GenBank mRNA | AF262055.1 | - | 4 - 5 | 440 | 87 | |
AK077167.1 | - | 4 - 5 | 445 | 87 | ||
AK146832.1 | - | 4 - 5 | 496 | 87 | ||
AU080225.1 | - | 4 - 5 | 511 | 87 | ||
BC052155.1 | - | 4 - 5 | 506 | 6335959 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |