Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161714.1 | NP_001155186.1 | 10 - 11 | 1600 | 87 | |
NM_001161715.1 | NP_001155187.1 | 10 - 11 | 1699 | 87 | ||
NM_019984.3 | NP_064368.3 | 10 - 11 | 1648 | 87 | ||
GenBank mRNA | AF186373.1 | - | 10 - 11 | 1591 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |