Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253679.1 | NP_001240608.1 | 8 - 9 | 1349 | 110 | |
NM_001253680.1 | NP_001240609.1 | 7 - 8 | 1367 | 110 | ||
NM_011405.4 | NP_035535.2 | 9 - 10 | 1506 | 110 | ||
GenBank mRNA | AJ012754.1 | - | 7 - 8 | 1357 | 110 | |
AJ130943.1 | - | 8 - 9 | 1349 | 110 | ||
AK150950.1 | - | 9 - 10 | 1506 | 110 | ||
AK151718.1 | - | 9 - 10 | 1506 | 110 | ||
AK152637.1 | - | 7 - 8 | 1272 | 110 | ||
AK170851.1 | - | 7 - 8 | 1278 | 110 | ||
BC014709.1 | - | 7 - 8 | 1355 | 4482816 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |