Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028946.3 | NP_083222.2 | 6 - 7 | 1206 | 59 | |
GenBank mRNA | AK016917.1 | - | 6 - 7 | 1186 | 59 | |
AK029525.1 | - | 6 - 7 | 1207 | 59 | ||
BC025002.1 | - | 5 - 6 | 1128 | 59 | ||
EU846100.1 | - | 5 - 6 | 737 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |