Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029256.3 | NP_083532.1 | 3 - 4 | 306 | 97 | |
NM_183293.1 | NP_899116.1 | 3 - 4 | 306 | 97 | ||
NR_111924.1 | 3 - 4 | 306 | 97 | |||
GenBank mRNA | AK016080.1 | - | 3 - 4 | 308 | 97 | |
AK016114.1 | - | 3 - 4 | 307 | 97 | ||
AK016535.1 | - | 3 - 4 | 293 | 97 | ||
BC120721.1 | - | 3 - 4 | 280 | 40129644 | 97 | |
BC120723.1 | - | 3 - 4 | 280 | 40129646 | 97 | |
BC145034.1 | - | 3 - 4 | 280 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |