Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pspc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025682.3 | NP_079958.3 | 5 - 6 | 1208 | 73 | |
GenBank mRNA | AK011272.1 | - | 5 - 6 | 1205 | 73 | |
AK150823.1 | - | 5 - 6 | 1209 | 73 | ||
AK160722.1 | - | 5 - 6 | 1209 | 73 | ||
BC026772.1 | - | 5 - 6 | 1187 | 3711631 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |