Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010194.2 | NP_034324.2 | 6 - 7 | 922 | 72 | |
GenBank mRNA | AK004587.1 | - | 6 - 7 | 914 | 72 | |
AK143639.1 | - | 6 - 7 | 872 | 72 | ||
AK170418.1 | - | 6 - 7 | 863 | 72 | ||
BC100306.1 | - | 5 - 6 | 1020 | 72 | ||
BC129919.1 | - | 5 - 6 | 858 | 40111855 | 72 | |
X12616.1 | - | 5 - 6 | 862 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |