Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201638.2 | NP_964000.2 | 6 - 7 | 665 | 133 | |
GenBank mRNA | AK083169.1 | - | 6 - 7 | 607 | 133 | |
AK089949.1 | - | 6 - 7 | 611 | 133 | ||
AK129409.1 | - | 623 | 133 | |||
AK146881.1 | - | 6 - 7 | 654 | 133 | ||
AK153810.1 | - | 6 - 7 | 661 | 133 | ||
BC052204.1 | - | 6 - 7 | 623 | 30007932 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |