Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027193.2 | NP_081469.2 | 2 - 3 | 252 | 135 | |
GenBank mRNA | AK010475.1 | - | 2 - 3 | 242 | 135 | |
AK034652.1 | - | 262 | 135 | |||
AK045127.1 | - | 2 - 3 | 272 | 135 | ||
AK045548.1 | - | 2 - 3 | 242 | 135 | ||
AK046024.1 | - | 2 - 3 | 242 | 135 | ||
AK046753.1 | - | 2 - 3 | 270 | 135 | ||
BC060372.1 | - | 2 - 3 | 252 | 6391546 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |