Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100030308.mm.1 |
NM_145514.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030308.mm.1 |
NM_145514.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030308.mm.1 |
TC0100003683.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030307.mm.1 |
NM_145514.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030307.mm.1 |
NM_145514.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030307.mm.1 |
TC0100003683.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030311.mm.1 |
NM_145514.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030311.mm.1 |
NM_145514.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030311.mm.1 |
TC0100003683.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030309.mm.1 |
NM_145514.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030309.mm.1 |
NM_145514.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030309.mm.1 |
TC0100003683.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030313.mm.1 |
NM_145514.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030313.mm.1 |
NM_145514.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030313.mm.1 |
TC0100003683.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030310.mm.1 |
NM_145514.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030310.mm.1 |
NM_145514.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030310.mm.1 |
TC0100003683.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100030314.mm.1 |
NM_145514.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030314.mm.1 |
NM_145514.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100030314.mm.1 |
TC0100003683.mm.1 |
MTA TC |