Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010513.2 | NP_034643.2 | 14 - 15 | 3490 | 75 | |
GenBank mRNA | AF056187.1 | - | 14 - 15 | 2888 | 75 | |
AK137220.1 | - | 9 - 10 | 1619 | 75 | ||
AK155884.1 | - | 13 - 14 | 2386 | 75 | ||
BC138868.1 | - | 14 - 15 | 2920 | 8861890 | 75 | |
BC138869.1 | - | 14 - 15 | 2920 | 8861891 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |