Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243046.1 | NP_001229975.1 | 5 - 6 | 595 | 73 | |
NM_001243047.1 | NP_001229976.1 | 2 - 3 | 374 | 73 | ||
NM_011807.3 | NP_035937.2 | 14 - 15 | 1850 | 73 | ||
GenBank mRNA | AF388675.1 | - | 14 - 15 | 1629 | 73 | |
AK039754.1 | - | 2 - 3 | 239 | 73 | ||
CB520190.1 | - | 2 - 3 | 374 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |