Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ticrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029835.1 | NP_084111.1 | 19 - 20 | 3453 | 131 | |
GenBank mRNA | AK051639.1 | - | 19 - 20 | 3416 | 131 | |
AK054492.1 | - | 19 - 20 | 3420 | 131 | ||
BC158103.1 | - | 19 - 20 | 3417 | 9088180 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |