Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ticrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029835.1 | NP_084111.1 | 20 - 21 | 5585 | 60 | |
GenBank mRNA | AK033104.1 | - | 1 - 2 | 1131 | 60 | |
BC058634.1 | - | 1 - 2 | 797 | 60 | ||
BC158103.1 | - | 20 - 21 | 5549 | 9088180 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |