Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sv2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109753.1 | NP_001103223.1 | 2 - 3 | 1015 | 65 | |
NM_153579.4 | NP_705807.2 | 2 - 3 | 1222 | 65 | ||
GenBank mRNA | AK029736.1 | - | 3 - 4 | 1047 | 65 | |
AK043830.1 | - | 2 - 3 | 942 | 65 | ||
AK122359.1 | - | 1 - 2 | 167 | 65 | ||
AK139478.1 | - | 2 - 3 | 917 | 65 | ||
AK140104.1 | - | 2 - 3 | 935 | 65 | ||
AK147261.1 | - | 2 - 3 | 1009 | 65 | ||
AK159028.1 | - | 2 - 3 | 920 | 65 | ||
BC060224.1 | - | 1 - 2 | 606 | 30356990 | 65 | |
BC079568.1 | - | 2 - 3 | 1222 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |