Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sv2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109753.1 | NP_001103223.1 | 7 - 8 | 1677 | 69 | |
NM_153579.4 | NP_705807.2 | 7 - 8 | 1884 | 69 | ||
GenBank mRNA | AK029736.1 | - | 8 - 9 | 1709 | 69 | |
AK043830.1 | - | 7 - 8 | 1604 | 69 | ||
AK122359.1 | - | 6 - 7 | 829 | 69 | ||
AK139478.1 | - | 7 - 8 | 1579 | 69 | ||
AK140104.1 | - | 7 - 8 | 1597 | 69 | ||
AK147261.1 | - | 7 - 8 | 1671 | 69 | ||
AK159028.1 | - | 7 - 8 | 1582 | 69 | ||
BC060224.1 | - | 6 - 7 | 1268 | 30356990 | 69 | |
BC079568.1 | - | 7 - 8 | 1884 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |