Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3glb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282042.1 | NP_001268971.1 | 9 - 10 | 1349 | 57 | |
GenBank mRNA | AK158415.1 | - | 9 - 10 | 1125 | 57 | |
DQ205686.1 | - | 9 - 10 | 1337 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis 3' Most Mus musculus Probe spans exons |