Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146191.3 | NP_666303.3 | 21 - 22 | 3374 | 95 | |
GenBank mRNA | AK051264.1 | - | 21 - 22 | 3362 | 95 | |
AK129441.1 | - | 18 - 19 | 2787 | 95 | ||
AK147469.1 | - | 21 - 22 | 3360 | 95 | ||
BC072664.1 | - | 21 - 22 | 3323 | 30543489 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |