Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033877.4 | NP_001029049.2 | 3 - 4 | 716 | 57 | |
GenBank mRNA | AK053131.1 | - | 3 - 4 | 717 | 57 | |
BC141170.1 | - | 3 - 4 | 668 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |