Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setdb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081024.1 | NP_001074493.1 | 7 - 8 | 1511 | 116 | |
GenBank mRNA | BC157926.1 | - | 6 - 7 | 1333 | 9088099 | 116 |
BC172021.1 | - | 6 - 7 | 1333 | 9054307 | 116 | |
EU155090.1 | - | 6 - 7 | 1325 | 116 | ||
EU155091.1 | - | 6 - 7 | 1325 | 116 | ||
EU155092.1 | - | 6 - 7 | 1343 | 116 | ||
EU155093.1 | - | 5 - 6 | 1259 | 116 | ||
EU155094.1 | - | 5 - 6 | 1259 | 116 | ||
EU155096.1 | - | 5 - 6 | 982 | 116 | ||
EU155097.1 | - | 7 - 8 | 1384 | 116 | ||
EU155099.1 | - | 6 - 7 | 1343 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |