Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048175.2 | NP_001041640.1 | 1 - 2 | 203 | 128 | |
NM_010082.2 | NP_034212.1 | 1 - 2 | 203 | 128 | ||
NM_183366.3 | NP_899222.1 | 1 - 2 | 203 | 128 | ||
NR_102399.1 | 1 - 2 | 203 | 128 | |||
GenBank mRNA | AF153350.1 | - | 1 - 2 | 110 | 128 | |
AF163290.1 | - | 1 - 2 | 203 | 128 | ||
AF163291.1 | - | 1 - 2 | 203 | 128 | ||
AF163292.1 | - | 1 - 2 | 203 | 128 | ||
AF163293.1 | - | 1 - 2 | 203 | 128 | ||
AK036800.1 | - | 1 - 2 | 143 | 128 | ||
AK052479.1 | - | 1 - 2 | 94 | 128 | ||
BC058782.1 | - | 1 - 2 | 78 | 6310763 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus 1700 Probe spans exons |