Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spert | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164141.1 | NP_001157613.1 | 2 - 3 | 227 | 80 | |
GenBank mRNA | AY092416.1 | - | 1 - 2 | 211 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons |