Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200000431.mm.1 |
NM_028376.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000431.mm.1 |
NM_028376.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000431.mm.1 |
TC1200000032.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000434.mm.1 |
NM_028376.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000434.mm.1 |
NM_028376.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000434.mm.1 |
TC1200000032.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000429.mm.1 |
NM_028376.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000429.mm.1 |
NM_028376.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000429.mm.1 |
TC1200000032.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000433.mm.1 |
NM_028376.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000433.mm.1 |
NM_028376.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000433.mm.1 |
TC1200000032.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000430.mm.1 |
NM_028376.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000430.mm.1 |
NM_028376.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000430.mm.1 |
TC1200000032.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000432.mm.1 |
NM_028376.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000432.mm.1 |
NM_028376.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000432.mm.1 |
TC1200000032.mm.1 |
MTA TC |