Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291184.1 | NP_001278113.1 | 10 - 11 | 1417 | 68 | |
NM_011048.1 | NP_035178.1 | 10 - 11 | 1417 | 68 | ||
GenBank mRNA | BC037450.1 | - | 8 - 9 | 1022 | 68 | |
D50060.1 | - | 10 - 11 | 1340 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |