Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chsy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081163.1 | NP_001074632.1 | 1 - 2 | 715 | 78 | |
GenBank mRNA | AK129255.1 | - | 1 - 2 | 715 | 78 | |
BC128567.1 | - | 2 - 3 | 162 | 78 | ||
BC153827.1 | - | 2 - 3 | 187 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |