Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dleu2 | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF380423.1 | - | LIT3422 DLEU2 |
4 - 5 | 456 | 123 | |
AF380425.1 | - | LIT3421 DLEU2 |
4 - 5 | 386 | 123 | ||
AK080165.1 | - | 4 - 5 | 282 | 123 | |||
AK084358.1 | - | 4 - 5 | 435 | 123 | |||
AK165901.1 | - | 4 - 5 | 469 | 123 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 15a | Mir15a |
Mirn15a, mir-15a, mmu-mir-15a |
Sense | Chr. 14 - 61632027 - 61632110 on NCBI Build GRCm38 | |
Downstream |
predicted gene, 27017 |
Gm27017 | 47.6kb | Anti-sense | Chr. 14 - 61680039 - 61686642 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |