Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cysltr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162412.1 | NP_001155884.1 | 3 - 4 | 257 | 150 | |
NM_133720.3 | NP_598481.2 | 4 - 5 | 314 | 207 | ||
GenBank mRNA | AF331658.1 | - | 4 - 5 | 273 | 207 | |
AK008997.1 | - | 3 - 4 | 258 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mus musculus Probe spans exons |