Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ripk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164107.1 | NP_001157579.1 | 8 - 9 | 1455 | 80 | |
NM_001164108.1 | NP_001157580.1 | 8 - 9 | 1553 | 80 | ||
NM_019955.2 | NP_064339.2 | 8 - 9 | 1472 | 80 | ||
GenBank mRNA | AF178953.1 | - | 8 - 9 | 1401 | 80 | |
AK012177.1 | - | 8 - 9 | 1445 | 80 | ||
AK154505.1 | - | 8 - 9 | 1465 | 80 | ||
BC029210.1 | - | 8 - 9 | 1423 | 3590770 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |