Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tatdn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163421.1 | NP_001156893.1 | 3 - 4 | 232 | 76 | |
NM_001163422.1 | NP_001156894.1 | 3 - 4 | 232 | 76 | ||
NM_026895.1 | NP_081171.1 | 3 - 4 | 232 | 76 | ||
GenBank mRNA | AK005193.1 | - | 3 - 4 | 203 | 76 | |
AK156071.1 | - | 3 - 4 | 183 | 76 | ||
BC061248.1 | - | 3 - 4 | 204 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |