Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Depdc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172092.1 | NP_001165563.1 | 2 - 3 | 406 | 104 | |
NM_001172093.1 | NP_001165564.1 | 2 - 3 | 406 | 104 | ||
NM_029523.3 | NP_083799.2 | 2 - 3 | 406 | 104 | ||
GenBank mRNA | AK050456.1 | - | 2 - 3 | 343 | 104 | |
AK088719.1 | - | 2 - 3 | 382 | 104 | ||
AK146458.1 | - | 2 - 3 | 377 | 104 | ||
BB857645.1 | - | 2 - 3 | 406 | 104 | ||
BC023934.1 | - | 2 - 3 | 399 | 5342167 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |