Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rarb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289760.1 | NP_001276689.1 | 7 - 8 | 1365 | 107 | |
NM_001289761.1 | NP_001276690.1 | 6 - 7 | 1284 | 107 | ||
NM_001289762.1 | NP_001276691.1 | 5 - 6 | 894 | 107 | ||
NM_011243.2 | NP_035373.1 | 5 - 6 | 1248 | 107 | ||
GenBank mRNA | AK078471.1 | - | 5 - 6 | 960 | 107 | |
BC076597.1 | - | 5 - 6 | 1244 | 30608242 | 107 | |
S56660.1 | - | 5 - 6 | 1144 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Nuclear Receptor Mammalian Gene Collection Mus musculus Probe spans exons |