Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm35066 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK035404.1 | - | 1 - 2 | 56 | 120 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 16233 | Gm16233 | OTTMUSG00000031903 | -18.3kb | Sense | Chr. 3 - 144947622 - 144953789 on NCBI Build GRCm38 |
Downstream | chloride channel calcium activated 1 | Clca1 |
Clca2, Clca3, gob-5, gob5 |
23.8kb | Anti-sense | Chr. 3 - 145004537 - 145032776 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons |