Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153564.2 | NP_705792.2 | 4 - 5 | 489 | 98 | |
GenBank mRNA | AF422243.1 | - | 4 - 5 | 471 | 98 | |
AK030414.1 | - | 4 - 5 | 489 | 98 | ||
AK087083.1 | - | 577 | 98 | |||
BC058555.1 | - | 4 - 5 | 523 | 5293575 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |