Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc45a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133952.2 | NP_598713.2 | 6 - 7 | 786 | 68 | |
GenBank mRNA | AK028912.1 | - | 6 - 7 | 772 | 68 | |
AK047292.1 | - | 6 - 7 | 733 | 68 | ||
AK047476.1 | - | 6 - 7 | 733 | 68 | ||
AK088034.1 | - | 6 - 7 | 738 | 68 | ||
AK166809.1 | - | 6 - 7 | 769 | 68 | ||
BC004717.1 | - | 6 - 7 | 775 | 3582116 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |