Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdccag8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029756.3 | NP_084032.1 | 10 - 11 | 1555 | 94 | |
GenBank mRNA | AK012392.1 | - | 10 - 11 | 1343 | 94 | |
AK132316.1 | - | 10 - 11 | 1351 | 94 | ||
AK133622.1 | - | 10 - 11 | 1299 | 94 | ||
BC016244.1 | - | 11 - 12 | 1450 | 4507575 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |