Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smyd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027188.3 | NP_081464.1 | 5 - 6 | 588 | 71 | |
GenBank mRNA | AK010447.1 | - | 5 - 6 | 622 | 71 | |
AK043988.1 | - | 5 - 6 | 598 | 71 | ||
AK163057.1 | - | 5 - 6 | 605 | 71 | ||
AK167875.1 | - | 5 - 6 | 603 | 71 | ||
BC019663.1 | - | 5 - 6 | 590 | 71 | ||
BC038482.1 | - | 5 - 6 | 469 | 71 | ||
BC052431.1 | - | 5 - 6 | 588 | 6837012 | 71 | |
BC061485.1 | - | 5 - 6 | 571 | 5370924 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |